EMAIL THIS PAGE TO A FRIEND
Biological trace element research
Nephrotoxicity and neurotoxicity in humans from organogermanium compounds and germanium dioxide.
PMID 1726409
Abstract
There is no known biological requirement for germanium (Ge), germanates, or any organogermanium compound. Ge deficiency has not been demonstrated in any animal. The estimated average dietary intake of Ge in humans is 1.5 mg/d. Ge is widely distributed in edible foods, all of which, with few exceptions, contain less than 5 ppm Ge, since higher levels are toxic to most plants. Ingestion of Ge compounds has been shown to produce toxic effects in experimental animals. In recent years inorganic germanium salts and novel organogermanium compounds, such as carboxyethyl germanium sesquioxide (Ge-132) and lactate-citrate-germanate (Ge lactate citrate) have been sold as "nutritional supplements" in some countries for their purported immunomodulatory effects or as health-producing elixirs, resulting in intakes of Ge significantly exceeding the estimated average dietary intake. Since 1982, there have been 18 reported cases of acute renal dysfunction or failure, including two deaths, linked to oral intake of Ge elixirs containing germanium dioxide (GeO2) or Ge-132. In these cases, biopsies show vacuolar degeneration in renal tubular epithelial cells, without proteinuria or hematuria, in the absence of glomerular changes. Serum creatinine levels have been well above 400 mumol/L in such patients. In 17 of 18 cases, accumulated elemental Ge intakes reportedly ranged between 16 to 328 g over a 4-36 mo period, or between 100 to 2000 times the average estimated dietary intake for human. In surviving patients, renal function improved after discontinuation of Ge supplementation. However, in no case was recovery complete. One organogermanium compound, an azaspiran organogermanium compound, 2-aza-8-germanspiro[4,5] decane-2-propamine-8,8-diethyl-N,N-dimethyl dichloride (spirogermanium), has been found to cause both neurotoxicity and pulmonary toxicity in phase I and II studies examining its chemotherapeutic potential as an antitumor drug in the treatment of various malignancies. In cancer patients given the drug spirogermanium, 40% experienced marked, yet transient neurotoxicity. Two patients suffered from pulmonary toxicity. Results of phases I and II human cancer trials for spirogermanium have not been favorable, with the exception of moderate benefits for three types of malignancies. It is recommended that patients exposed to long-term (greater than 3 mo) Ge supplementation at levels well above the estimated daily intake be medically supervised and monitored for potential renal-, pulmonary- or neurotoxicity. Further study regarding the mechanism of Ge-induced nephrotoxicity in human is warranted.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF58081451
Germanium, sheet, 50x50mm, thickness 3.0mm, polycrystalline, 99.999%
Ge
GF99720647
Germanium, sheet, 50x50mm, thickness 1.0mm, polycrystalline, 99.999%
Ge
GF31896366
Germanium, sheet, 7x24mm, thickness 1.0mm, polycrystalline, 99.999%
Ge
GF46650090
Germanium, disks, 15mm, thickness 1.0mm, polycrystalline, 100%
Ge
GF90754858
Germanium, disks, 20mm, thickness 1.0mm, single crystal, 100%
Ge
GF77697680
Germanium, microfoil, 25x25mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%
Ge
GF58880739
Germanium, microfoil, 25x25mm, thinness 0.5μm, specific density 333μg/cm2, 6 micron aluminum permanent support, 100%
Ge
GF12210624
Germanium, microfoil, 50x50mm, thinness 0.25μm, specific density 166.3μg/cm2, 6 micron aluminum permanent support, 100%
Ge
GF57527219
Germanium, rod, 25mm, diameter 2.0mm, polycrystalline, n-type, 99.999%
Ge
GF65029417
Germanium, rod, 25mm, diameter 5mm, polycrystalline, n-type, 99.999%
Ge
GF14446346
Germanium, rod, 50mm, diameter 5mm, polycrystalline, n-type, 99.999%
Ge
GF04605780
Germanium, rod, 6mm, diameter 6.0mm, single crystal, 100%
Ge
GF09210417
Germanium, sheet, 10x10mm, thickness 0.10mm, polycrystalline, 99.999%
Ge
GF00633664
Germanium, sheet, 10x10mm, thickness 0.25mm, polycrystalline, 99.999%
Ge
GF09629859
Germanium, sheet, 10x10mm, thickness 0.5mm, single crystal, -111, 100%
Ge
GF45727241
Germanium, sheet, 10x10mm, thickness 0.6mm, single crystal, -111, 100%
Ge
GF44621580
Germanium, sheet, 25x25mm, thickness 1.0mm, polycrystalline, 99.999%
Ge
GF44356978
Germanium, sheet, 25x25mm, thickness 1.0mm, single crystal, 99.999%
Ge
GF20558022
Germanium, sheet, 25x25mm, thickness 3.0mm, polycrystalline, 99.999%
Ge
GF49319815
Germanium, sheet, 4x4mm, thickness 0.05mm, polycrystalline, 99.999%
Ge
GF73619107
Germanium, sheet, 50x50mm, thickness 0.5mm, single crystal, 99.999%
Ge
GF37208369
Germanium, sheet, 6x6mm, thickness 1.0mm, polycrystalline, 99.999%
Ge