EMAIL THIS PAGE TO A FRIEND
Steroids
CoMSIA and docking study of rhenium based estrogen receptor ligand analogs.
PMID 17280694
Abstract
OPLS all atom force field parameters were developed in order to model a diverse set of novel rhenium based estrogen receptor ligands whose relative binding affinities (RBA) to the estrogen receptor alpha isoform (ERalpha) with respect to 17beta-estradiol were available. The binding properties of these novel rhenium based organometallic complexes were studied with a combination of Comparative Molecular Similarity Indices Analysis (CoMSIA) and docking. A total of 29 estrogen receptor ligands consisting of 11 rhenium complexes and 18 organic ligands were docked inside the ligand-binding domain (LBD) of ERalpha utilizing the program Gold. The top ranked pose was used to construct CoMSIA models from a training set of 22 of the estrogen receptor ligands which were selected at random. In addition scoring functions from the docking runs and the polar volume (PV) were also studied to investigate their ability to predict RBA ERalpha. A partial least-squares analysis consisting of the CoMSIA steric, electrostatic and hydrophobic indices together with the polar volume proved sufficiently predictive having a correlation coefficient, r(2), of 0.94 and a cross-validated correlation coefficient, q(2), utilizing the leave-one-out method of 0.68. Analysis of the scoring functions from Gold showed particularly poor correlation to RBA ERalpha which did not improve when the rhenium complexes were extracted to leave the organic ligands. The combined CoMSIA and polar volume model ranked correctly the ligands in order of increasing RBA ERalpha, illustrating the utility of this method as a prescreening tool in the development of novel rhenium based estrogen receptor ligands.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
459984
Rhenium, foil, thickness 1.0 mm, 99.98% trace metals basis
Re
267317
Rhenium, foil, thickness 0.25 mm, 99.98% trace metals basis
Re
267309
Rhenium, foil, thickness 0.1 mm, 99.98% trace metals basis
Re
204188
Rhenium, powder, 99.995% trace metals basis
Re
267279
Rhenium, powder, −100 mesh, ≥99.9% trace metals basis
Re
357138
Rhenium, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Re
GF57863931
Rhenium, wire reel, 0.1m, diameter 0.25mm, annealed, 99.97%
Re
GF60560513
Rhenium, wire reel, 0.1m, diameter 0.178mm, annealed, 99.97%
Re
GF90374441
Rhenium, wire reel, 0.1m, diameter 0.3mm, annealed, 99.97%
Re
GF77806367
Rhenium, wire reel, 0.1m, diameter 0.5mm, annealed, 99.97%
Re
GF63991929
Rhenium, wire reel, 0.1m, diameter 1.0mm, annealed, 99.97%
Re
GF14519156
Rhenium, wire reel, 0.2m, diameter 0.076mm, annealed, 99.97%
Re
GF59830139
Rhenium, wire reel, 0.2m, diameter 0.10mm, annealed, 99.97%
Re
GF96591875
Rhenium, wire reel, 0.2m, diameter 0.125mm, annealed, 99.97%
Re
GF02183217
Rhenium, wire reel, 0.2m, diameter 0.15mm, annealed, 99.97%
Re
GF70064658
Rhenium, wire reel, 0.2m, diameter 0.178mm, annealed, 99.97%
Re
GF32991486
Rhenium, wire reel, 0.2m, diameter 0.25mm, annealed, 99.97%
Re
GF48358435
Rhenium, wire reel, 0.2m, diameter 0.2mm, annealed, 99.97%
Re
GF96805884
Rhenium, wire reel, 0.2m, diameter 0.3mm, annealed, 99.97%
Re
GF25796010
Rhenium, wire reel, 0.2m, diameter 0.5mm, annealed, 99.97%
Re
GF98990050
Rhenium, wire reel, 0.2m, diameter 1.0mm, annealed, 99.97%
Re
GF14894163
Rhenium, wire reel, 0.5m, diameter 0.076mm, annealed, 99.97%
Re
GF78766745
Rhenium, wire reel, 0.5m, diameter 0.10mm, annealed, 99.97%
Re
GF73984797
Rhenium, wire reel, 0.5m, diameter 0.125mm, annealed, 99.97%
Re
GF72901324
Rhenium, wire reel, 0.5m, diameter 0.15mm, annealed, 99.97%
Re
GF58081945
Rhenium, wire reel, 0.5m, diameter 0.178mm, annealed, 99.97%
Re
GF23936104
Rhenium, wire reel, 0.5m, diameter 0.25mm, annealed, 99.97%
Re
GF44804325
Rhenium, wire reel, 0.5m, diameter 0.2mm, annealed, 99.97%
Re
GF00735353
Rhenium, wire reel, 0.5m, diameter 0.3mm, annealed, 99.97%
Re
GF24502115
Rhenium, wire reel, 0.5m, diameter 0.5mm, annealed, 99.97%
Re