EMAIL THIS PAGE TO A FRIEND
Operative dentistry
Effect of curing mode on microtensile bond strength to dentin of two dual-cured adhesive systems in combination with resin luting cements for indirect restorations.
PMID 17288327
Abstract
This study evaluated the microtensile bond strength (microTBS) of dual-cured adhesive systems when the different components were either light activated or left in the uncured state prior to cementation of an indirect composite restoration. Occlusal dentin surfaces of 40 human third molars were flattened. The teeth were randomly assigned to 8 groups (n = 5) according to the dual-cured systems (bonding agents/resin cements) and curing modes: All Bond 2/Duolink (AB2-BISCO Inc) and Optibond Solo Plus Dual Cure/Nexus 2 (SOLO-Kerr). Resin cements were applied to pre-cured resin composite discs (2 mm thick/Z-250/3M ESPE), which were fixed to dentin surfaces containing adhesive resin in either cured (LP) or uncured states (SP). The restored teeth were light activated according to the manufacturers' instructions (LRC-XL3000/3M ESPE) or allowed to self-cure (SRC). The restored teeth were water-stored at 37 degrees C for 24 hours. They were then both mesial-distally and buccal-lingually sectioned to obtain bonded specimens (1.2 mm2). Each specimen was tested in tension at a crosshead speed of 0.6 mm/minute until failure. Data (MPa (SD)) were analyzed by two-way ANOVA and Tukey's post-hoc test (p < .05). AB2/SP exhibited higher microTBS than AB2/LP (p = .00001); however, no significant differences were noted between SOLO/LP and SOLO/SP. Results suggested that dual-cured adhesive systems were as strong or even stronger when they were left in the uncured state prior to indirect resin composite cementation.