EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Diclofenac solubility: independent determination of the intrinsic solubility of three crystal forms.
PMID 17298048
Abstract
The solubility in water of diclofenac ({2-[(2,6-dichlorophenyl)amino]phenyl}acetic acid), a potent nonsteroidal anti-inflammatory drug, has been investigated. The various solid forms have been characterized by thermogravimetric analysis, differential scanning calorimetry, and X-ray diffraction. The commercially available form of diclofenac is the anhydrous sodium salt. This was recrystallized from ethanol and precipitated as a hydrate containing four diclofenac anions, four sodium cations, and nineteen water molecules per unit cell. This crystal structure is similar to but different from an earlier report of the structure. Crystals of the acid form of diclofenac were anhydrous and corresponded to an earlier crystal structure. Separate solubility measurements on all three of these solid forms of diclofenac gave consistent results for the intrinsic solubility. The aqueous solubility values reported in the literature for diclofenac are spread over a large range, with a factor of 100 separating the largest and the smallest. Our value is at the smaller end of this range. It is the only one supported by three independent procedures and rigorous characterization of the solid forms. The experimental conditions were precisely controlled.