EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Total synthesis of papulacandin D.
PMID 17305346
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
144207
Chlorodimethylsilane, 98%
C2H7ClSi