EMAIL THIS PAGE TO A FRIEND
BMC molecular biology
Interleukin-4 inhibition of interleukin-1-induced expression of matrix metalloproteinase-3 (MMP-3) is independent of lipoxygenase and PPARgamma activation in human gingival fibroblasts.
PMID 17319946
Abstract
Interleukin 4 (IL-4) has been shown to suppress interleukin-1 (IL-1) induced expression of matrix metalloproteinase-3 (MMP-3) in human synovial and gingival fibroblasts, but the mechanism of suppression has not been determined. Activators of peroxisome proliferator-activated receptor-gamma (PPARgamma) have been shown to inhibit cytokine induced expression of MMPs in other cell types, and IL-4 has been shown to activate PPARgamma by stimulating production of ligands through the lipoxygenase pathway. It has been suggested that PPARgamma may inhibit expression of MMPs by competing with transcription factor AP-1 for binding to a putative composite binding element in the promoters. The objective of this study was to determine whether the suppressive effects of IL-4 on the IL-1 induced expression of MMP-3 involve activation of lipoxygenase and/or PPARgamma. Western blotting revealed the presence of PPARgamma in nuclear extract of HGF. IL-1 induced binding of nuclear extract to the putative composite PPRE/AP-1 site was diminished in the presence of pioglitazone, but there was no evidence of any change in the composition of the retarded complexes, and no evidence of PPARgamma binding to this site. Nordihydroguaiaretic acid (NDGA), a non-selective lipoxygenase inhibitor, and MK886, a specific inhibitor of 5-lipoxygenase, induced MMP-3 expression synergistically with IL-1. However IL-4 was still able to inhibit MMP-3 expression in the presence of NDGA or MK886 and IL-1. Activation of PPARgamma with pioglitazone not only failed to inhibit IL-1 induced expression of MMP-3 mRNA, but rather super-induced MMP-3 in the presence of IL-1. PPARgamma antagonist GW9662 failed to abolish the suppressive effects of IL-4. Another PPARgamma activator, 15-deoxy-Delta12,14prostaglandin J2 (15dPGJ2), also super-induced MMP-3 mRNA, and this was due at least in part to increased transcription. IL-4 suppression of IL-1-induced MMP-3 expression in HGF is independent of lipoxygenase activity and activation of PPARgamma. Super-induction of MMP-3 by pioglitazone may have important implications for patients using pioglitazone to treat type II diabetes in the presence of chronic inflammation.