EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Expression and purification of an active form of the Mycobacterium leprae DNA gyrase and its inhibition by quinolones.
PMID 17325221
Abstract
Mycobacterium leprae, the causative agent of leprosy, is noncultivable in vitro; therefore, evaluation of antibiotic activity against M. leprae relies mainly upon the mouse footpad system, which requires at least 12 months before the results become available. We have developed an in vitro assay for studying the activities of quinolones against the DNA gyrase of M. leprae. We overexpressed in Escherichia coli the M. leprae GyrA and GyrB subunits separately as His-tagged proteins by using a pET plasmid carrying the gyrA and gyrB genes. The soluble 97.5-kDa GyrA and 74.5-kDa GyrB subunits were purified by nickel chelate chromatography and were reconstituted as an enzyme with DNA supercoiling activity. Based on the drug concentrations that inhibited DNA supercoiling by 50% or that induced DNA cleavage by 25%, the 13 quinolones tested clustered into three groups. Analysis of the quinolone structure-activity relationship demonstrates that the most active quinolones against M. leprae DNA gyrase share the following structural features: a substituted carbon at position 8, a cyclopropyl substituent at N-1, a fluorine at C-6, and a substituent ring at C-7. We conclude that the assays based on DNA supercoiling inhibition and drug-induced DNA cleavage on purified M. leprae DNA gyrase are rapid, efficient, and safe methods for the screening of quinolone derivatives with potential in vivo activities against M. leprae.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O0877
Oxolinic acid, quinolone antibiotic
C13H11NO5
56968
Sparfloxacin, ≥98.0% (HPLC)
C19H22F2N4O3