EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Multiple functions of the IKK-related kinase IKKepsilon in interferon-mediated antiviral immunity.
PMID 17332413
Abstract
IKKepsilon is an IKK (inhibitor of nuclear factor kappaBkinase)-related kinase implicated in virus induction of interferon-beta (IFNbeta). We report that, although mice lacking IKKepsilon produce normal amounts of IFNbeta, they are hypersusceptible to viral infection because of a defect in the IFN signaling pathway. Specifically, a subset of type I IFN-stimulated genes are not activated in the absence of IKKepsilon because the interferon-stimulated gene factor 3 complex (ISGF3) does not bind to promoter elements of the affected genes. We demonstrate that IKKepsilon is activated by IFNbeta and that IKKepsilon directly phosphorylates signal transducer and activator of transcription 1 (STAT1), a component of ISGF3. We conclude that IKKepsilon plays a critical role in the IFN-inducible antiviral transcriptional response.