EMAIL THIS PAGE TO A FRIEND
World journal of urology
Holmium laser for stone management.
PMID 17340157
Abstract
The efficiency and safety profile of the holmium laser has made this tool a versatile multi-purpose instrument for use in the endoscopic treatment of a wide variety of urologic disorders, in particular urinary calculi. Herein we review holmium laser physics, current endourologic laser lithotripsy applications, and the performance of new low power holmium laser devices.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
457957
Holmium, chips, 99.9% trace metals basis
Ho
693693
Holmium, foil, thickness 1 mm, 99.9% trace rare earth metals basis
Ho
755974
Holmium, dendritic pieces, purified by distillation, 99.99% trace rare earth metals basis
Ho
GF92551259
Holmium, rod, 50mm, diameter 3.8mm, 99%
Ho
GF12230154
Holmium, rod, 50mm, diameter 6.35mm, cast, 99%
Ho
GF39809863
Holmium, wire reel, 100mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF22997188
Holmium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Ho
GF59427513
Holmium, wire reel, 200mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF74648223
Holmium, wire reel, 200mm, diameter 1.0mm, hard, 99.9%
Ho
GF88732693
Holmium, wire reel, 50mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF13376785
Holmium, rod, 200mm, diameter 3.8mm, 99%
Ho
GF28769541
Holmium, rod, 100mm, diameter 6.35mm, cast, 99%
Ho
GF68907011
Holmium, rod, 100mm, diameter 3.8mm, 99%
Ho
GF31028543
Holmium, powder, 5g, max. particle size 500 micron, 99.9%
Ho
GF78567654
Holmium, powder, 10g, max. particle size 500 micron, 99.9%
Ho
GF29991673
Holmium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Ho
GF26625067
Holmium, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99%
Ho
GF42613006
Holmium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99%
Ho
GF60938635
Holmium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Ho
GF34764666
Holmium, foil, not light tested, 50x50mm, thickness 0.0125mm, as rolled, 99%
Ho
GF78609640
Holmium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Ho
GF49265253
Holmium, foil, not light tested, 25x25mm, thickness 0.0125mm, as rolled, 99%
Ho
GF07383977
Holmium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Ho
GF85502640
Holmium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Ho
GF19613684
Holmium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Ho
GF38519376
Holmium, foil, not light tested, 100x100mm, thickness 0.0125mm, as rolled, 99%
Ho