EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and pharmacological evaluation of novel conformationally constrained homologues of glutamic acid.
PMID 17346858
Abstract
Twelve novel conformationally constrained homologues of glutamic acid have been synthesized and pharmacologically characterized at ionotropic glutamate receptors (iGluRs). Synthesis of the target compounds involved 1,3-dipolar cycloaddition of nitrile oxides to suitable dipolarophiles. The structure to the compounds has been assigned by (1)H NMR and, in the case of derivatives (+/-)-4a, (+/-)-4b, (+/-)-5a, and (+/-)-5b, by means of an X-ray crystallographic analysis carried out on intermediate (+/-)-12a. The synthesized amino acids were found to be without affinity (K(i)/IC(50)>100microM) for iGluRs with the exception of compounds (+/-)-4b and (+/-)-5b, which showed a modest affinity for NMDA receptors (K(i)=34 and 13microM, respectively). The results indicate that the increased conformational constraints introduced by the cyclopropane ring and the spiro-attached proline ring are both detrimental to the pharmacological activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8054
D(−)-2-Amino-5-phosphonopentanoic acid, NMDA receptor antagonist
C5H12NO5P