EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Analogues of acifran: agonists of the high and low affinity niacin receptors, GPR109a and GPR109b.
PMID 17358052
Abstract
Recently identified GPCRs, GPR109a and GPR109b, the high and low affinity receptors for niacin, may represent good targets for the development of HDL elevating drugs for the treatment of atherosclerosis. Acifran, an agonist of both receptors, has been tested in human subjects, yet until recently very few analogs had been reported. We describe a series of acifran analogs prepared using newly developed synthetic pathways and evaluated as agonists for GPR109a and GPR109b, resulting in identification of compounds with improved activity at these receptors.