EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Zinc stannate (Zn2SnO4) dye-sensitized solar cells.
PMID 17371032
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
391433
tert-Butylamine, purified by redistillation, ≥99.5%
C4H11N