EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Endothelin-2/vasoactive intestinal contractor via ROCK regulates transglutaminase 1 on differentiation of mouse keratinocytes.
PMID 17418814
Abstract
We previously found that endothelin-2/vasoactive intestinal contractor (ET-2/VIC) greatly increased in mouse epidermis after birth. In the present study, we evaluated whether ET-2/VIC expression was associated with the calcium-induced differentiation of cultured mouse keratinocytes. The differentiation induction was revealed by morphological change, cornified envelope (CE) formation, and involucrin and transglutaminase 1 (TG 1) expressions. ET-2/VIC gene expression and peptide production subsequently increased in the induction of the differentiation. We also found that Y-27632, a Rho-associated coiled-coil forming protein serine/threonine kinase (ROCK) inhibitor, suppressed up-regulation of ET-2/VIC gene expression, the induction of morphological change, the CE formation, and TG 1 expression, but not involucrin expression. These results indicate new three findings, (1) ET-2/VIC expression increases and has potential as a differentiation marker, (2) ET-2/VIC expression is mediated by ROCK, and (3) the ROCK regulated TG 1 expression, on the calcium-induced differentiation of mouse keratinocytes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SCP0259 Vasoactive Intestinal Constrictor / β Endothelin mouse, ≥96% (HPLC)
C116H161N27O32S4