EMAIL THIS PAGE TO A FRIEND
BJU international
Peri-operative complications of holmium laser enucleation of the prostate: experience in the first 280 patients, and a review of literature.
PMID 17419697
Abstract
To evaluate, in a prospective study, the complications in 280 patients undergoing holmium laser enucleation of the prostate (HoLEP) at our institution, and to review previous reports to determine the overall incidence and types of various complications, and analyse their causes and means of prevention. We analysed the patients' demographic, peri-operative and follow-up data, and the complications during and after surgery. HoLEP was completed successfully in 268 patients (95.7%); eight required conversion to transurethral resection of the prostate (TURP) during the initial experience. The morcellation device and laser malfunctioned in two patients each. A blood transfusion was required during HoLEP in one patient; other complications included capsular perforation (9.6%), superficial bladder mucosal injury (3.9%) and ureteric orifice injury (2.1%). A blood transfusion was needed after HoLEP in 1.4% of patients and cystoscopy with clot evacuation in 0.7%. Transient urinary incontinence was the commonest complication after HoLEP, in 10.7% of patients, but recovered spontaneously in all except two (0.7%). Other rare complications were re-catheterization (3.9%), urinary tract infection (3.2%), epididymitis (0.7%), meatal and submeatal stenosis (2.5%), bulbar urethral stricture (2.1%), bladder neck contracture (0.35%) and myocardial infarction (0.35%). There was a low incidence of complications with HoLEP; most were minor and easily managed. Our results are comparable with those published previously, and establish HoLEP as safe and reproducible procedure. While gaining experience, HoLEP can be converted to TURP with no harm to the patient.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
457957
Holmium, chips, 99.9% trace metals basis
Ho
693693
Holmium, foil, thickness 1 mm, 99.9% trace rare earth metals basis
Ho
755974
Holmium, dendritic pieces, purified by distillation, 99.99% trace rare earth metals basis
Ho
GF92551259
Holmium, rod, 50mm, diameter 3.8mm, 99%
Ho
GF12230154
Holmium, rod, 50mm, diameter 6.35mm, cast, 99%
Ho
GF39809863
Holmium, wire reel, 100mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF22997188
Holmium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Ho
GF59427513
Holmium, wire reel, 200mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF74648223
Holmium, wire reel, 200mm, diameter 1.0mm, hard, 99.9%
Ho
GF88732693
Holmium, wire reel, 50mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF13376785
Holmium, rod, 200mm, diameter 3.8mm, 99%
Ho
GF28769541
Holmium, rod, 100mm, diameter 6.35mm, cast, 99%
Ho
GF68907011
Holmium, rod, 100mm, diameter 3.8mm, 99%
Ho
GF31028543
Holmium, powder, 5g, max. particle size 500 micron, 99.9%
Ho
GF78567654
Holmium, powder, 10g, max. particle size 500 micron, 99.9%
Ho
GF29991673
Holmium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Ho
GF26625067
Holmium, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99%
Ho
GF42613006
Holmium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99%
Ho
GF60938635
Holmium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Ho
GF34764666
Holmium, foil, not light tested, 50x50mm, thickness 0.0125mm, as rolled, 99%
Ho
GF78609640
Holmium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Ho
GF49265253
Holmium, foil, not light tested, 25x25mm, thickness 0.0125mm, as rolled, 99%
Ho
GF07383977
Holmium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Ho
GF85502640
Holmium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Ho
GF19613684
Holmium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Ho
GF38519376
Holmium, foil, not light tested, 100x100mm, thickness 0.0125mm, as rolled, 99%
Ho