EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Syntheses and antibacterial activity of phendioxy substituted cyclic enediynes.
PMID 17433674
Abstract
Syntheses and antibacterial activity of 13-membered 1,3-phendioxy substituted cyclic enediynes are reported. The compounds were screened against gram-positive and gram-negative strains and some of the compounds exhibit potent antibacterial activity.