EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
2,3,5-Substituted tetrahydrofurans as cancer chemopreventives. Part 1: synthesis and anti-cancer activities of 5-hydroxymethyl-2,3-diaryl-tetrahydro-furan-3-ols.
PMID 17446077
Abstract
The allylation of appropriate benzoin in presence of indium metal followed by m-CPBA mediated cyclization gave 5-hydroxymethyl-2,3-diaryl-tetrahydro-furan-3-ols. Investigations on 59 human tumor cell lines of these compounds identify four compounds exhibiting significant growth inhibition of tumor cells at particular cell lines. Compound 12 is very specific toward CCRF-CEM and SR cell lines of leukemia.