EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Technetium-99m-labeling and synthesis of thymidine analogs: potential candidates for tumor imaging.
PMID 17449246
Abstract
The technetium-99m-labeling and synthesis of a series of thymidine analogs were studied. The target molecules were obtained by using 6-hydrazinopyridine-3-carboxylic acid (HYNIC) as a bifunctional coupling agent and using N-(2-hydroxy-1,1-bis(hydroxymethyl)ethyl)glycine (tricine) and ethylenediamine-N,N'-diacetic acid (EDDA) as coligands. The effects of different spacers between thymidine analog with HYNIC on radiochemical yield were also studied.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
100013
4,4′-Dimethoxytrityl chloride, 95%
C21H19ClO2
158186
Ethylenediamine-N,N′-diacetic acid, ≥98%
C6H12N2O4