EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Vibrio splendidus as the source of plasmid-mediated QnrS-like quinolone resistance determinants.
PMID 17452482
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56968
Sparfloxacin, ≥98.0% (HPLC)
C19H22F2N4O3