EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Combined therapies in a murine model of blastoschizomycosis.
PMID 17452487
Abstract
In a murine model of blastoschizomycosis, amphotericin B combined with micafungin, flucytosine or voriconazole did not improve the efficacy of fluconazole. However, such combinations can constitute therapeutic options for those cases where fluconazole fails.