EMAIL THIS PAGE TO A FRIEND
Masui. The Japanese journal of anesthesiology
[Description of anesthesia through the xenon binding response in a protein model under a pressure-of 2,000-anesthetic binding with low specificity produces resistance to pressure with the volume increase].
PMID 17458133
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00472
Xenon, ≥99.995%
Xe