EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Cucurbitane glycosides from the fruits of Siraitia gros venorii and their inhibitory effects on Epstein-Barr virus activation.
PMID 17477572
Abstract
Six new cucurbitane glycosides, mogroside II B (2), 11-deoxymogroside III (4), 7-oxomogroside II E (5), 7-oxomogroside V (6), 11-oxomogroside II A1 (7), and 11-oxomogroside IV A (8), and two known but new naturally occurring cucurbitane glycosides, mogroside II A1 (1) and mogroside III A2 (3), were isolated from an ethanol extract of the fruits of Siraitia grosvenorii. Upon evaluation of compounds 1-8 for inhibitory effects against the Epstein-Barr virus early antigen (EBV-EA) activation induced by 12-O-tetradecanoylphorbol-13-acetate (TPA), all compounds exhibited inhibitory effects with IC50 values of 346-400 mol ratio/32 pmol TPA. In addition, compounds 1-8 showed weak inhibitory effects on activation of (+/-)-(E)-methyl-2-[(E)-hydroxyimino]-5-nitro-6-methoxy-3-hexemide (NOR 1), a nitric oxide (NO) donor.