EMAIL THIS PAGE TO A FRIEND
Organic letters
Recoverable PEG-supported copper catalyst for highly stereocontrolled nitroaldol condensation.
PMID 17480085
Abstract
A new poly(ethylene glycol)-modified DAT2-Cu(OAc)2 complex smoothly catalyzes a base-free nitroaldol condensation in a highly enantioselective manner (ee up to 93%) also in reagent-grade solvent and in the presence of air. Effective recovery and recycling (up to five runs) of supported catalysts are documented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
126292
3,5-Dimethoxybenzaldehyde, 98%
C9H10O3