EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Design, docking, and synthesis of novel indeno[1,2-c]isoquinolines for the development of antitumor agents as topoisomerase I inhibitors.
PMID 17498951
Abstract
An intramolecular radical cyclization reaction of 4-bromo-3-arylisoquinolines 11a-c allowed the efficient synthesis of 11-methylindenoisoquinolines 2a-c. 5-(2-Aminoethylamino)indeno[1,2-c]isoquinolin-11-one 4 was also prepared in the convenient manner. The synthesized compounds were tested in vitro for cytotoxicity and DNA-topoisomerase 1 (top 1) inhibitory activity. The dramatic enhancement of top 1 inhibitory activity of 4 was explained by a docking study using the FlexX program.