EMAIL THIS PAGE TO A FRIEND
The Journal of nutrition
Orotic acid excretion and arginine metabolism.
PMID 17513443
Abstract
The urinary excretion of orotic acid, an intermediate in the pyrimidine biosynthetic pathway, is markedly increased in many inborn errors of the urea cycle and in a number of other disorders involving arginine metabolism. Carbamoyl phosphate, which accumulates within hepatic mitochondria in patients with ornithine transcarbamoylase deficiency, can diffuse to the cytosol and enter the pyrimidine pathway, resulting in greatly increased orotic acid production and excretion. This orotic aciduria also occurs in inborn errors of the mitochondrial ornithine/citrulline transporter, arginase, argininosuccinate synthetase, and argininosuccinate lyase. Increased orotic acid excretion is also found in a number of hypoargininemic states, such as lysinuric protein intolerance. However, orotic aciduria should not be used uncritically as an index of arginine deficiency because it is found in patients with arginase deficiency who exhibit hyperargininemia. Increased orotic acid excretion can also arise as a result of impairments of pyrimidine synthesis, whether brought about by a genetic defect (e.g., in UMP synthase) or by drugs that inhibit the terminal part of the pathway (e.g., allopurinol or 6-azauridine). When used appropriately, measurement of urinary orotic acid is a valuable tool for the study of many derangements of arginine metabolism, including arginine depletion, and to assess the efficacy of therapies used to replete this amino acid.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1882
6-Azauridine
C8H11N3O6
O2750
Orotic acid, ≥98% (titration), anhydrous
C5H4N2O4