EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and biological activity of new anti-inflammatory compounds containing the 1,4-benzodioxine and/or pyrrole system.
PMID 17517512
Abstract
A series of substituted derivatives containing the 1,4-benzodioxine or pyrrole nucleus are described. All the newly synthesized compounds were examined for their in vitro and in vivo anti-inflammatory activity. Several derivatives, including (S)-2, 14 and 17, showed more anti-inflammatory activity in vivo in these assays (rat paw oedema induced by carrageenan) than the known classical anti-inflammatory agent ibuprofen, whereas other compounds like 1 were equipotent to ibuprofen. Compound 17 was the most outstanding derivative because of its remarkable in vivo anti-inflammatory activity. In this paper, we examine and discuss the structure-activity relationships and anti-inflammatory activities of these compounds.