EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
First occurrence of an Escherichia coli clinical isolate producing the VIM-1/VIM-2 hybrid metallo-beta-lactamase VIM-12.
PMID 17517838