EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Changes in qnr prevalence and fluoroquinolone resistance in clinical isolates of Klebsiella pneumoniae and Enterobacter spp. collected from 1990 to 2005.
PMID 17526754
Abstract
Clinical isolates of Klebsiella pneumoniae and Enterobacter spp. collected from 1990 through 2005 at a tertiary care center were studied for qnr genes. Isolates bearing these genes emerged in the mid-1990s, coinciding with the time of a rapid increase in fluoroquinolone resistance. Sixty percent of these isolates were ciprofloxacin susceptible by CLSI breakpoints.