EMAIL THIS PAGE TO A FRIEND
Kardiologiia
[Clinical pharmacology of nisoldipine, a new calcium antagonist].
PMID 1753601
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N0165
Nisoldipine, ≥98% (HPLC)
C20H24N2O6