EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V
Dry powder inhalation of hemin to induce heme oxygenase expression in the lung.
PMID 17537624
Abstract
The purpose of this study was to formulate hemin as a powder for inhalation and to show proof of concept of heme oxygenase 1 (HO-1) expression in the lungs of mice by inhalation of hemin. Hemin was spray dried from a neutralized sodium hydroxide solution. The particle size distribution of the powder was between 1 and 5 microm. Dispersion from the Twincer dry powder inhaler showed a fine particle fraction (<5 microm) of 36%. A specially designed aerosol box based on the Twincer-inhaler was used for a proof of concept study of HO-1 induction by inhalation of hemin in mice. The aerosol in the exposure chamber of the aerosol box remained aerosolized up to 5 min. A rhodamin B containing aerosol was used to show that the aerosol box gave deposition over the entire lung indicating the suitability of the model. Additionally, inhalation of hemin showed a dose dependent increase in HO-1 protein expression in the lungs. In conclusion, hemin was successfully formulated as a powder for inhalation and the inhalation model allowed controlled HO-1 expression in the lungs of mice. Future studies investigating the utility of inhaled hemin in treating disease states are warranted.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51280
Hemin, BioXtra, from Porcine, ≥97.0% (HPLC)
C34H32ClFeN4O4