EMAIL THIS PAGE TO A FRIEND
The Journal of antimicrobial chemotherapy
Decreased affinity of mosaic-structure recombinant penicillin-binding protein 2 for oral cephalosporins in Neisseria gonorrhoeae.
PMID 17540669
Abstract
In Neisseria gonorrhoeae, the mosaic structure of penicillin-binding protein 2 (PBP 2), composed of fragments of PBP 2 from Neisseria cinerea and Neisseria perflava, was significantly associated with decreased susceptibility to cephalosporins, particularly oral cephalosporins. The aim of this study was to determine the affinity of mosaic PBP 2 for cephalosporins in N. gonorrhoeae. Two types of non-mosaic PBP 2 from the type strain of N. gonorrhoeae (ATCC 19424) and a clinical strain (GU01-29), as well as the mosaic PBP 2 from a clinical strain (GU01-89), were expressed in insect cells, and recombinant PBP 2s were purified. ATCC 19424 and GU01-29 were susceptible to cephalosporins. GU01-89 showed decreased susceptibility to cephalosporins. Bindings of fluorescent penicillin to PBP 2 were characterized by the Scatchard plot analysis. The affinity of the recombinant PBP 2s for cefdinir, cefixime and ceftriaxone was determined by PBP 2 competition assays with fluorescent penicillin. The K(d) value of mosaic PBP 2 for fluorescent penicillin was higher than that of non-mosaic PBP 2s. The affinity of mosaic PBP 2 for cefdinir or cefixime was lower than that of the non-mosaic PBP 2s. The affinity of the mosaic PBP 2 for ceftriaxone was not changed, compared with that of the non-mosaic PBP 2s. Other mechanisms may be involved in clinical isolates with decreased susceptibility to cephalosporins, but this study suggests that the decreased affinity of mosaic-structure recombinant PBP 2 for oral cephalosporins may contribute to decreased susceptibility to these antibiotics in N. gonorrhoeae.