EMAIL THIS PAGE TO A FRIEND
Organic letters
Development of a catalytic aromatic decarboxylation reaction.
PMID 17542594
Abstract
A palladium-catalyzed aromatic decarboxylation reaction has been developed. With electron-rich aromatic acids, the reaction proceeds efficiently under fairly mild conditions in good yields. The method was useful with complex functionalized substrates containing hindered carboxylic acids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
299685
Palladium(II) trifluoroacetate, 97%
C4F6O4Pd