EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Exomethylene pyranonucleosides: efficient synthesis and biological evaluation of 1-(2,3,4-trideoxy-2-methylene-beta-d-glycero-hex-3-enopyranosyl)thymine.
PMID 17555969
Abstract
A new series of unsaturated pyranonucleosides with an exocyclic methylene group and thymine as heterocyclic base have been designed and synthesized. d-Galactose (1) was readily transformed in three steps into the corresponding 1-(beta-d-galactopyranosyl)thymine (2). Selective protection of the primary hydroxyl group of 2 with a t-butyldimethylsilyl (TBDMS) group, followed by specific acetalation, and oxidation gave 1-(6-O-t-butyldimethylsilyl-3,4-O-isopropylidene-beta-d-lyxo-hexopyranosyl-2-ulose)thymine (5). Wittig reaction of the ketonucleoside 5, deprotection and tritylation of the 6'-hydroxyl group gave 1-(2-deoxy-2-methylene-6-O-trityl-beta-d-lyxo-hexopyranosyl)thymine (9). Exomethylene pyranonucleoside 9 was converted to the olefinic derivative 10, which after detritylation afforded the title compound 1-(2,3,4-trideoxy-2-methylene-beta-d-glycero-hex-3-enopyranosyl)thymine (11). These novel synthesized compounds were evaluated for antiviral activity against rotaviral infection and cytotoxicity in colon cancer. As compared to AZT, compounds 1-(2-deoxy-2-methylene-beta-d-lyxo-hexopyranosyl)thymine (7) and 1-(beta-d-lyxo-hexopyranosyl-2-ulose)thymine (8) showed to be more efficient, in rotavirus infections and in treatment of colon cancer.