EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Dual bioactivity of resveratrol fatty alcohols: differentiation of neural stem cells and modulation of neuroinflammation.
PMID 17560107
Abstract
The synthesis of resveratrol fatty alcohols (RFAs), a new class of small molecules presenting strong potential for the treatment of neurological diseases, is described. RFAs, hybrid compounds combining the resveratrol nucleus and omega-alkanol side chains, are able to modulate neuroinflammation and to induce differentiation of neural stem cells into mature neurons. Acting on neuroprotection and neuroregeneration, RFAs represent an innovative approach for the treatment or cure of neuropathies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
526088
Diethyl 4-methoxybenzylphosphonate, ≥98%
C12H19O4P