EMAIL THIS PAGE TO A FRIEND
Rheumatology (Oxford, England)
Radiosynoviorthesis of medium-sized joints with rhenium-186-sulphide colloid: a review of the literature.
PMID 17596287
Abstract
Hypertrophy and inflammation of the synovium with various underlying pathologies - such as rheumatoid arthritis, osteoarthritis, haemophilia and spondyloarthropathy - can be treated successfully by radiosynoviorthesis (RSO). For medium-sized joints (shoulder, elbow, wrist, hip and ankle), the radionuclide of choice is rhenium-186. We review the evidence for the efficacy of this local, relatively non-invasive therapy and evaluate its benefits and risks. We conclude good evidence of rhenium-186 RSO in rheumatoid arthritis and haemophilic arthropathy. In the remaining pathologies, up to now, the therapeutic efficacy has not been confirmed by today's most stringent criteria for clinical studies. The available data support rhenium-186 RSO as a suitable second-line treatment for patients in whom other therapies (including locally injected corticoids) have failed, as long as proper attention is paid to correct administration - including post-treatment immobilization and the co-administration of corticoids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
459984
Rhenium, foil, thickness 1.0 mm, 99.98% trace metals basis
Re
267317
Rhenium, foil, thickness 0.25 mm, 99.98% trace metals basis
Re
267309
Rhenium, foil, thickness 0.1 mm, 99.98% trace metals basis
Re
204188
Rhenium, powder, 99.995% trace metals basis
Re
267279
Rhenium, powder, −100 mesh, ≥99.9% trace metals basis
Re
357138
Rhenium, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Re
GF57863931
Rhenium, wire reel, 0.1m, diameter 0.25mm, annealed, 99.97%
Re
GF60560513
Rhenium, wire reel, 0.1m, diameter 0.178mm, annealed, 99.97%
Re
GF90374441
Rhenium, wire reel, 0.1m, diameter 0.3mm, annealed, 99.97%
Re
GF77806367
Rhenium, wire reel, 0.1m, diameter 0.5mm, annealed, 99.97%
Re
GF63991929
Rhenium, wire reel, 0.1m, diameter 1.0mm, annealed, 99.97%
Re
GF14519156
Rhenium, wire reel, 0.2m, diameter 0.076mm, annealed, 99.97%
Re
GF59830139
Rhenium, wire reel, 0.2m, diameter 0.10mm, annealed, 99.97%
Re
GF96591875
Rhenium, wire reel, 0.2m, diameter 0.125mm, annealed, 99.97%
Re
GF02183217
Rhenium, wire reel, 0.2m, diameter 0.15mm, annealed, 99.97%
Re
GF70064658
Rhenium, wire reel, 0.2m, diameter 0.178mm, annealed, 99.97%
Re
GF32991486
Rhenium, wire reel, 0.2m, diameter 0.25mm, annealed, 99.97%
Re
GF48358435
Rhenium, wire reel, 0.2m, diameter 0.2mm, annealed, 99.97%
Re
GF96805884
Rhenium, wire reel, 0.2m, diameter 0.3mm, annealed, 99.97%
Re
GF25796010
Rhenium, wire reel, 0.2m, diameter 0.5mm, annealed, 99.97%
Re
GF98990050
Rhenium, wire reel, 0.2m, diameter 1.0mm, annealed, 99.97%
Re
GF14894163
Rhenium, wire reel, 0.5m, diameter 0.076mm, annealed, 99.97%
Re
GF78766745
Rhenium, wire reel, 0.5m, diameter 0.10mm, annealed, 99.97%
Re
GF73984797
Rhenium, wire reel, 0.5m, diameter 0.125mm, annealed, 99.97%
Re
GF72901324
Rhenium, wire reel, 0.5m, diameter 0.15mm, annealed, 99.97%
Re
GF58081945
Rhenium, wire reel, 0.5m, diameter 0.178mm, annealed, 99.97%
Re
GF23936104
Rhenium, wire reel, 0.5m, diameter 0.25mm, annealed, 99.97%
Re
GF44804325
Rhenium, wire reel, 0.5m, diameter 0.2mm, annealed, 99.97%
Re
GF00735353
Rhenium, wire reel, 0.5m, diameter 0.3mm, annealed, 99.97%
Re
GF24502115
Rhenium, wire reel, 0.5m, diameter 0.5mm, annealed, 99.97%
Re