EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Regiochemistry of copper(I)-mediated cyclization reactions of halo-dienamides.
PMID 17602591
Abstract
Reaction of 2-substituted dienamides with catalytic amounts of copper halide/tripyridylamine (TPA) furnishes either 5-exo or 6-endo products with the outcome dependent upon the radical initiating unit. Reaction of 3-substituted dienamides produces beta-lactams via a 4-exo cyclization with termination of the reaction occurring via either halogen atom transfer, trapping with oxygen, elimination, or radical-radical coupling depending upon the diene.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
252271
α-Bromoisobutyryl bromide, 98%
C4H6Br2O