EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Bioactive peroxides as potential therapeutic agents.
PMID 17618015
Abstract
Present review describes research on more than 280 natural anticancer agents isolated from terrestrial and marine sources and synthetic biologically active peroxides. Intensive searches for new classes of pharmacologically potent agents produced by terrestrial and marine organisms have resulted in the discovery of dozens of compounds possessing high cytotoxic, antibacterial, antimalarial, and other activities as an important source of leads for drug discovery.