EMAIL THIS PAGE TO A FRIEND
Journal of translational medicine
Haplotype of gene Nedd4 binding protein 2 associated with sporadic nasopharyngeal carcinoma in the Southern Chinese population.
PMID 17626640
Abstract
Bcl-3 as an oncoprotein is overexpressed in nasopharyngeal carcinoma (NPC). Nedd4 binding protein 2 (N4BP2), which is located in the NPC susceptibility locus, is a Bcl-3 binding protein. This study is aimed to explore the association between N4BP2 genetic polymorphism and the risk of NPC. We performed a hospital-based case-control study, including 531 sporadic NPC and 480 cancer-free control subjects from southern China. PCR-sequencing was carried out on Exons, promoter region and nearby introns of the N4BP2 gene. The expression pattern of N4BP2 and Bcl-3 was also analyzed. We observed a statistically significant difference in haplotype blocks ATTA and GTTG between cases and controls. In addition, three novel SNPs were identified, two of which were in exons (loc123-e3l-snp2, position 39868005, A/G, Met171Val; RS17511668-SNP2, position 39926432, G/A, Glu118Lys), and one was in the intron6 (RS794001-SNP1, position 39944127, T/G). Moreover, N4BP2 was at higher levels in a majority of tumor tissues examined, relative to paired normal tissues. These data suggest that haplotype blocks ATTA and GTTG of N4BP2 is correlation with the risk of sporadic nasopharyngeal carcinoma in the Southern Chinese population and N4BP2 has a potential role in the development of NPC.