EMAIL THIS PAGE TO A FRIEND
Journal of peptide science : an official publication of the European Peptide Society
Synthesis of lipophilic 2-oxoamides based on gamma-aminobutyric and delta-aminovaleric analogues and their activity against phospholipase A2.
PMID 17631670
Abstract
A variety of lipophilic 2-oxoamides based on gamma-aminobutyric and delta-aminovaleric analogues were synthesized. 2-oxoamides containing a tetrazole, a thioethyl or a thioacetyl group are weak inhibitors of GIVA cPLA(2), while derivatives containing a methyl tetrazole, a diethyl phosphonate or a thioethyl group are weak inhibitors of GV sPLA(2).