EMAIL THIS PAGE TO A FRIEND
BMC immunology
TSLP is involved in expansion of early thymocyte progenitors.
PMID 17640367
Abstract
Thymic stromal derived lymphopoietin (TSLP) is preferentially and highly expressed in the thymus, but its function in T cell development is not clear. We report here that TSLP, independently or in combination with IL-7, enhances thymopoiesis in the murine fetal thymic organ culture (FTOC) model. Furthermore, TSLP preferentially increases the number and proliferation of the (DN1 and DN2) pro-T progenitor cells, and FTOC lobes from TSLP receptor-null mice show a decreased number of these cells. Finally, DN1-DN2 cells expanded with TSLP in vitro are functional T progenitors that are able to differentiate into mature T cells in fetal or adult thymus organs. Together, these data suggest that TSLP plays an important role in expansion of thymocyte progenitors and may be of value for expanding T progenitor cells in vitro.