EMAIL THIS PAGE TO A FRIEND
Organic letters
Convenient synthesis of palladium nanoparticles and catalysis of Hiyama coupling reaction in water.
PMID 17676861
Abstract
An efficient synthesis of Pd nanoparticles in water has been developed using a Fischer carbene complex of tungsten as the reductant and PEG as the capping agent. The colloidal palladium (1 mol %) efficiently catalyzes Hiyama cross-coupling reactions performed in air. Excellent yields of products were obtained with a wide range of substrates. Catalytic activity and stability of the nanoparticles were found to be inversely correlated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
241431
Tungsten hexacarbonyl, 97%
C6O6W