EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Optimization of the dosage of flucytosine in combination with amphotericin B for disseminated candidiasis: a pharmacodynamic rationale for reduced dosing.
PMID 17682101
Abstract
Amphotericin B and flucytosine (5FC) have an additive effect when used for disseminated candidiasis. Here, we bridge the results of an experimental pharmacodynamic study to humans and demonstrate that a 5FC dosage of 25 mg/kg of body weight/day in four divided doses in combination with amphotericin B produces near-maximal effect.