EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Design and evaluation of naphthol- and carbazole-containing fluorescent sigma ligands as potential probes for receptor binding studies.
PMID 17713896
Abstract
Some 3,3-dimethyl-1-(3-naphthylpropyl)piperidine and 1-cyclohexyl-4-(3-naphthylpropyl)piperazine derivatives, structurally containing naphthol as a fluorescent moiety, were prepared for being potentially used as fluorescent sigma ligands. Structurally related analogs were also prepared, where the naphthalene nucleus was replaced by the fluorescent carbazole moiety and chain length was varied. For all compounds the in vitro affinities toward sigma receptors and Delta8-Delta7 sterol isomerase site were measured, and the fluorescent properties were determined. Compound 19 gave the best results both for sigma receptor affinities (sigma1, Ki = 6.78 nM and sigma2, Ki = 26.4 nM) and fluorescence features; thus, it was chosen for in vitro saturation binding analysis at sigma receptors. The good results obtained in such assay suggested that the fluorescent compound 19 could be used instead of a radioligand in "green" binding assays.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P134
(−)-Pentazocine, ≥98% (HPLC)
C19H27NO