EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis, structure analysis, and antibacterial activity of some novel 10-substituted 2-(4-piperidyl/phenyl)-5,5-dioxo[1,2,4]triazolo[1,5-b][1,2,4]benzothiadiazine derivatives.
PMID 17719222
Abstract
A new series of 10-substituted 5,5-dioxo-5,10-dihydro[1,2,4]triazolo[1,5-b]-[1,2,4]benzothiadiazine arylsulfonamide derivatives (10a-j and 13a-f) was synthesized. The structures of these compounds were confirmed on the basis of spectral data, elemental analysis, X-ray analysis, and quantum chemical calculations. These compounds were evaluated for their efficacy as antibacterial agents against various Gram-positive and Gram-negative strains of bacteria. Amongst these compounds 10f and 10i were the most active compounds against Escherichia coli and 13e against E. coli as well as Bacillus subtilis. Moreover, other compounds also showed potent inhibitory activity in comparison to the standard drugs.