EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Fabrication and magnetic properties of arrays of metallic nanowires.
PMID 17731862
Abstract
Arrays of ferromagnetic nickel and cobalt nanowires have been fabricated by electrochemical deposition of the metals into templates with nanometer-sized pores prepared by nuclear track etching. These systems display distinctive characteristics because of their one-dimensional microstructure. The preferred magnetization direction is perpendicular to the film plane. Enhanced coercivities as high as 680 oersteds and remnant magnetization up to 90 percent have also been observed.