EMAIL THIS PAGE TO A FRIEND
Clinica chimica acta; international journal of clinical chemistry
Prenatal diagnosis of isovaleric acidemia by fast atom bombardment and tandem mass spectrometry.
PMID 1777996
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50405
2-Methylbutyryl-L-carnitine, ≥97.0% (TLC)
C12H23NO4