EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Indoloxypropanolamine analogues as 5-HT(1A) receptor antagonists.
PMID 17804228
Abstract
Analogues of pindolol, 1-(1H-indol-4-yloxy)-3-isopropylamino-propan-2-ol, were synthesized and evaluated as 5-HT(1A) receptor antagonists. The structural features required for optimal binding to the 5-HT1A receptor are as follows: S-2-propanol linker, 4-indoloxy substituent, and a large lipophilic cyclic amine substituent.