EMAIL THIS PAGE TO A FRIEND
Clinical reviews in allergy
Allergy to formaldehyde and ethylene-oxide.
PMID 1782627
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48838
Ethylene oxide solution, certified reference material, 50 mg/mL in methanol
C2H4O