EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Convenient synthesis of indeno[1,2-c]isoquinolines as constrained forms of 3-arylisoquinolines and docking study of a topoisomerase I inhibitor into DNA-topoisomerase I complex.
PMID 17827007
Abstract
11-hydroxyindeno[1,2-c]isoquinolines 12a-c were prepared as constrained forms of 3-arylisoquinolines through an intramolecular cyclization reaction. Among the synthesized compounds, the 11-(i)butoxy analog 15l displayed potent in vitro cytotoxicity against four different tumor cell lines as well as topoisomerase 1 inhibitory activity. A FlexX docking study was performed to explain the topoisomerase 1 activity of 15l.