EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Abscisic Acid Induces Formation of Floating Leaves in the Heterophyllous Aquatic Angiosperm Potamogeton nodosus.
PMID 17830317
Abstract
Potamogeton nodosus tubers produce floating-type instead of submersed-type leaves when exposed to 10(-5) molar synthetic abscisic acid. Abscisic acid-induced leaves have stomata on upper leaf surfaces and higher width/length ratios than controls. These effects are wholly or partially overcome by simultaneous exposure to abscisic acid combined with gibberellic acid, kinetin, or benzyladenine.