EMAIL THIS PAGE TO A FRIEND
Annals of surgery
THE TREATMENT OF ACID AND ALKALI BURNS: AN EXPERIMENTAL STUDY.
PMID 17865648
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
438073
Nitric acid, ACS reagent, 70%
HNO3